Stampa…

Anti-Poliovirus 1 Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB23658-100EA 0 EUR
BIRBORB23658-100
Anti-Poliovirus 1 Mouse Monoclonal Antibody
Anticorpi
Immunofluorescence: 1:100-1:400

Type: Primary
Antigen: Poliovirus 1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR