Stampa…

Anti-HSP-70, Geodia cydonium Mouse Monoclonal Antibody

Fornitore: Biorbyt
BIRBORB23653-100EA 0 EUR
BIRBORB23653-100
Anti-HSP-70, Geodia cydonium Mouse Monoclonal Antibody
Anticorpi
ELISA: 1:5000; Western Blot: 1:500; Immunofluorescence: 1:50

Type: Primary
Antigen: HSP-70, Geodia cydonium
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR