Stampa…
Anti-PAICS Rabbit Polyclonal Antibody
Fornitore: Abnova
ABNOH00010606-D01PEA 380 EUR
ABNOH00010606-D01P
Anti-PAICS Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: PAICS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human