Italy
Order Entry
Anti-Rac/Cdc42 guanine nucleotide exchange Factor (GEF) 6 Rabbit Polyclonal Antibody
  ABNOH00009459-D01P
 :  Abnova
undefined
Anti-Rac/Cdc42 guanine nucleotide exchange Factor (GEF) 6 Rabbit Polyclonal Antibody
  ABNOH00009459-D01P
 :  Abnova
 
364,00 €
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Rac/Cdc42 guanine nucleotide exchange Factor (GEF) 6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9459
  • Antigen synonyms:
    MRX46|PIXA|alpha-PIX|COOL2|alphaPIX|Cool-2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: Rac/Cdc42 guanine nucleotide exchange Factor (GEF) 6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human