Anti-DST Mouse Monoclonal Antibody [clone: 1D2]
ABNOH00000667-M02
: Abnova
- Antibody type:Primary
- Antigen name:dystonin
- Antigen symbol:DST
- Clonality:Monoclonal
- Clone:1D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:667
- Antigen synonyms:D6S1101|CATX15|BPAG1|CATX-15|MACF2|DMH|DT|EBSB2|HSAN6|BPA|BP240
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: DST
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human