Stampa…

Anti-FXYD3 Mouse Monoclonal Antibody [clone: OTI1F12]

Fornitore: OriGene
ORIGTA504726EA 616 EUR
ORIGTA504726
Anti-FXYD3 Mouse Monoclonal Antibody [clone: OTI1F12]
Anticorpi
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: FXYD3 (FXYD domain containing ion transport regulator 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR