Stampa…

Anti-ACSF2 Mouse Monoclonal Antibody [clone: OTI6E4]

Fornitore: OriGene
ORIGTA504711EA 621 EUR
ORIGTA504711
Anti-ACSF2 Mouse Monoclonal Antibody [clone: OTI6E4]
Anticorpi
Recommended Dilutions: Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACSF2 (acyl-CoA synthetase family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR