Stampa…

Anti-RUNX1T1 Mouse Monoclonal Antibody [clone: OTI1H1]

Fornitore: OriGene
ORIGTA504703EA 621 EUR
ORIGTA504703
Anti-RUNX1T1 Mouse Monoclonal Antibody [clone: OTI1H1]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: RUNX1T1 (runt-related transcription factor 1; translocated to, 1 (cyclin D-related))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR