Stampa…

Anti-TMEFF2 Mouse Monoclonal Antibody [clone: OTI3H8]

Fornitore: OriGene
ORIGTA504482EA 616 EUR
ORIGTA504482
Anti-TMEFF2 Mouse Monoclonal Antibody [clone: OTI3H8]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TMEFF2 (transmembrane protein with EGF-like and two follistatin-like domains 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR