Stampa…

Anti-CST3 Mouse Monoclonal Antibody [clone: OTI2C8]

Fornitore: OriGene
ORIGTA504442EA 616 EUR
ORIGTA504442
Anti-CST3 Mouse Monoclonal Antibody [clone: OTI2C8]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CST3 (cystatin C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR