Stampa…

Anti-ECI2 Mouse Monoclonal Antibody [clone: OTI2H10]

Fornitore: OriGene
ORIGTA504437EA 616 EUR
ORIGTA504437
Anti-ECI2 Mouse Monoclonal Antibody [clone: OTI2H10]
Anticorpi
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ECI2 (enoyl-CoA delta isomerase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR