Stampa…

Anti-SERPINA5 Mouse Monoclonal Antibody [clone: OTI1C4]

Fornitore: OriGene
ORIGTA504369EA 616 EUR
ORIGTA504369
Anti-SERPINA5 Mouse Monoclonal Antibody [clone: OTI1C4]
Anticorpi
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:100

Type: Primary
Antigen: SERPINA5 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR