Stampa…

Anti-EPN2 Mouse Monoclonal Antibody [clone: OTI1F5]

Fornitore: OriGene
ORIGTA504301EA 593 EUR
ORIGTA504301
Anti-EPN2 Mouse Monoclonal Antibody [clone: OTI1F5]
Anticorpi
  • Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: EPN2 (epsin 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR