Stampa…

Anti-CST4 Mouse Monoclonal Antibody [clone: OTI1C9]

Fornitore: OriGene
ORIGTA504094EA 593 EUR
ORIGTA504094
Anti-CST4 Mouse Monoclonal Antibody [clone: OTI1C9]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CST4 (cystatin S)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR