Stampa…

Anti-C1S Mouse Monoclonal Antibody [clone: OTI3B11]

Fornitore: OriGene
ORIGTA504080EA 593 EUR
ORIGTA504080
Anti-C1S Mouse Monoclonal Antibody [clone: OTI3B11]
Anticorpi
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: C1S (complement component 1, s subcomponent)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR