Stampa…

Anti-MTFMT Mouse Monoclonal Antibody [clone: OTI1E7]

Fornitore: OriGene
ORIGTA503565EA 616 EUR
ORIGTA503565
Anti-MTFMT Mouse Monoclonal Antibody [clone: OTI1E7]
Anticorpi
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150

Type: Primary
Antigen: MTFMT (mitochondrial methionyl-tRNA formyltransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR