Stampa…

Anti-PPARA Mouse Monoclonal Antibody [clone: OTI2G6]

Fornitore: OriGene
ORIGTA503545EA 616 EUR
ORIGTA503545
Anti-PPARA Mouse Monoclonal Antibody [clone: OTI2G6]
Anticorpi
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR