Stampa…

Anti-PMVK Mouse Monoclonal Antibody [clone: OTI2D1]

Fornitore: OriGene
ORIGTA503448EA 621 EUR
ORIGTA503448
Anti-PMVK Mouse Monoclonal Antibody [clone: OTI2D1]
Anticorpi
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: PMVK (phosphomevalonate kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR