Stampa…

Anti-ACY1 Mouse Monoclonal Antibody [clone: OTI2D3]

Fornitore: OriGene
ORIGTA503190EA 616 EUR
ORIGTA503190
Anti-ACY1 Mouse Monoclonal Antibody [clone: OTI2D3]
Anticorpi
Recommended Dilutions: Western Blot: 1:200-500; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: ACY1 (aminoacylase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR