Stampa…

Anti-EIF4E Mouse Monoclonal Antibody [clone: OTI9H1]

Fornitore: OriGene
ORIGTA503029EA 616 EUR
ORIGTA503029
Anti-EIF4E Mouse Monoclonal Antibody [clone: OTI9H1]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: EIF4E (eukaryotic translation initiation factor 4E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR