Stampa…

Anti-PDXK Mouse Monoclonal Antibody [clone: OTI5H5]

Fornitore: OriGene
ORIGTA502998EA 621 EUR
ORIGTA502998
Anti-PDXK Mouse Monoclonal Antibody [clone: OTI5H5]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PDXK (pyridoxal (pyridoxine, vitamin B6) kinase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR