Stampa…

Anti-DEF8 Mouse Monoclonal Antibody [clone: OTI2H4]

Fornitore: OriGene
ORIGTA502670EA 621 EUR
ORIGTA502670
Anti-DEF8 Mouse Monoclonal Antibody [clone: OTI2H4]
Anticorpi
Recommended Dilutions: Western Blot: 1:500; Flow Cytometry: 1:100

Type: Primary
Antigen: DEF8 (differentially expressed in FDCP 8 homolog (mouse))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR