Stampa…

Anti-SULT1C2 Mouse Monoclonal Antibody [clone: OTI6F2]

Fornitore: OriGene
ORIGTA502664EA 616 EUR
ORIGTA502664
Anti-SULT1C2 Mouse Monoclonal Antibody [clone: OTI6F2]
Anticorpi
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT1C2 (sulfotransferase family, cytosolic, 1C, member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR