Stampa…

Anti-VASP Mouse Monoclonal Antibody [clone: OTI4D6]

Fornitore: OriGene
ORIGTA502647EA 598 EUR
ORIGTA502647
Anti-VASP Mouse Monoclonal Antibody [clone: OTI4D6]
Anticorpi
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: VASP (vasodilator-stimulated phosphoprotein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR