Stampa…

Anti-MAGEB18 Mouse Monoclonal Antibody [clone: OTI1H2]

Fornitore: OriGene
ORIGTA502633EA 593 EUR
ORIGTA502633
Anti-MAGEB18 Mouse Monoclonal Antibody [clone: OTI1H2]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: MAGEB18 (melanoma antigen family B, 18)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR