Stampa…

Anti-SNTG1 Mouse Monoclonal Antibody [clone: OTI3G10]

Fornitore: OriGene
ORIGTA502224EA 621 EUR
ORIGTA502224
Anti-SNTG1 Mouse Monoclonal Antibody [clone: OTI3G10]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SNTG1 (syntrophin, gamma 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR