Stampa…

Anti-HP Mouse Monoclonal Antibody [clone: OTI1F9]

Fornitore: OriGene
ORIGTA501674EA 616 EUR
ORIGTA501674
Anti-HP Mouse Monoclonal Antibody [clone: OTI1F9]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: HP (haptoglobin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR