Stampa…

Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI3D4]

Fornitore: OriGene
ORIGTA501051EA 616 EUR
ORIGTA501051
Anti-PTPRE Mouse Monoclonal Antibody [clone: OTI3D4]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:50

Type: Primary
Antigen: PTPRE (protein tyrosine phosphatase, receptor type, E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR