Stampa…

Anti-IFT57 Mouse Monoclonal Antibody [clone: OTI5B6]

Fornitore: OriGene
ORIGTA500906EA 616 EUR
ORIGTA500906
Anti-IFT57 Mouse Monoclonal Antibody [clone: OTI5B6]
Anticorpi
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50

Type: Primary
Antigen: IFT57 (intraflagellar transport 57 homolog (Chlamydomonas))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR