Stampa…

Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI9H10]

Fornitore: OriGene
ORIGTA500704EA 621 EUR
ORIGTA500704
Anti-RNF144B Mouse Monoclonal Antibody [clone: OTI9H10]
Anticorpi
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: RNF144B (ring finger protein 144B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR