Stampa…

Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI5C3]

Fornitore: OriGene
ORIGTA500437EA 616 EUR
ORIGTA500437
Anti-NRBP1 Mouse Monoclonal Antibody [clone: OTI5C3]
Anticorpi
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NRBP1 (nuclear receptor binding protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR