Stampa…

Anti-LAMP1 Rabbit Polyclonal Antibody

Fornitore: OriGene
ORIGTA317793EA 809 EUR
ORIGTA317793
Anti-LAMP1 Rabbit Polyclonal Antibody
Anticorpi
Recommended Dilutions: Immunochemistry: 5 µg/ml

Type: Primary
Antigen: LAMP1 (lysosomal-associated membrane protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR