Stampa…

Anti-SLC1A5 Rabbit Polyclonal Antibody

Fornitore: OriGene
ORIGTA317316EA 680 EUR
ORIGTA317316
Anti-SLC1A5 Rabbit Polyclonal Antibody
Anticorpi
Recommended Dilutions: Immunochemistry: 2.5 µg/ml

Type: Primary
Antigen: SLC1A5 (solute carrier family 1 (neutral amino acid transporter), member 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR