Stampa…

Anti-OR5P2 Rabbit Polyclonal Antibody

Fornitore: OriGene
ORIGTA316044EA 564 EUR
ORIGTA316044
Anti-OR5P2 Rabbit Polyclonal Antibody
Anticorpi
Recommended Dilutions: Western Blot: 1:500-1:3000; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: OR5P2 (olfactory receptor, family 5, subfamily P, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR