Stampa…

Anti-AKAP11 Rabbit Polyclonal Antibody

Fornitore: OriGene
ORIGTA315160EA 564 EUR
ORIGTA315160
Anti-AKAP11 Rabbit Polyclonal Antibody
Anticorpi
Recommended Dilutions: Immunochemistry: 1:50-1:100; Immunofluorescence: 1:100-1:500; ELISA: 1:10000

Type: Primary
Antigen: AKAP11 (A kinase (PRKA) anchor protein 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR