Italy
Order Entry
Anti-FTO Goat Antibody
  ORIGTA305858
 :  OriGene
Anti-FTO Goat Antibody
  ORIGTA305858
 :  OriGene
 
785,00 €
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Fat Mass and Obesity-associated Protein
  • Antigen symbol:
    FTO
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
  • Sequence:
    QQKPDCRPYWEKDD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide. Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20C. Minimize freezing and thawing.
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 003-01?g/ml

Type: Primary
Antigen: FTO (fat mass and obesity associated)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: