Anti-PKR (PRKR) Rabbit Polyclonal Antibody
Catalog # ABGEAP8151B
Supplier: Abgent
Supplier Article Number: AP8151B
Local Article Number: ABGEAP8151B
Price
354,00 €
Log in for availability
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 2 alpha kinase 2
- Antigen symbol:EIF2AK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Gene ID:6625
- Antigen synonyms:double-stranded RNA-activated protein kinase|Interferon-induced|Interferon-inducible RNA-dependent protein kinase|EIF2AK2|P1/eIF-2A protein kinase|PKR|p68 kinase
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:HSGRDDTSRYDERPC
- Storage temperature:Cool
- Concentration:0,5 mg/ml
- Shipping temperature:Ice
- Immunogen:This SNRNP70 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 195-223 amino acids from the Central region of human SNRNP70.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PKR (PRKR)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse