Italy
Order Entry
Anti-AIMP2 Rabbit Polyclonal Antibody
  ABGEAP2861C
 :  Abgent
undefined
Anti-AIMP2 Rabbit Polyclonal Antibody
  ABGEAP2861C
 :  Abgent
 
354,00 €
   
  1 * 400 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Aminoacyl tRNA Synthetase Complex-interacting multifunctional Protein 2
  • Antigen symbol:
    AIMP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    AIMP2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: AIMP2 (aminoacyl tRNA synthetase complex-interacting multifunctional protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human