Anti-PML Rabbit Polyclonal Antibody
AVIVARP33048_P050
- Antibody type:Primary
- Antigen name:promyelocytic leukemia
- Antigen symbol:PML
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:5371
- Antigen synonyms:TRIM19|RNF71|PP8675|MYL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PML (promyelocytic leukemia)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: