Stampa…
Test tube racks
Fornitore: New Brunswick
NEWBM1289-0020EA 404 EUR
NEWBM1289-0020 NEWBM1289-0010 NEWBM1289-0040 NEWBM1289-0030 NEWBM1289-0060 NEWBM1289-0050 NEWBM1289-0006 NEWBM1289-0400 NEWBM1289-0004 NEWBM1289-0500 NEWBM1289-0005 NEWBM1289-0002 NEWBM1289-0200 NEWBM1289-0003 NEWBM1289-0300 NEWBM1289-0100 NEWBM1289-0001 444-4550
Test tube racks
Rack Rastrelliere per tubi
Stainless steel
Test tube racks contains inserts to retain tubes and prevent them from spinning in the rack. Finger-tight knobs for adjusting the angle and releasing the rack from the base plate for easy transport to the bench.