Stampa…
Eye showers, wall-mounted
Fornitore: asecos
MSGA26734EA 319 EUR
MSGA26734
Eye showers, wall-mounted
These eye showers feature a handle made of fibre glass reinforced polyamide, shower heads covered with rubber protection, leak-tested lids and dust caps. For wall installation.
  • Shower heads with large flushing area
  • 20° inclined towards the user
  • Arresting release button
  • Valve stays open after first use
  • Dust protection cap easily falls off during use
  • Flexible tube covered with stainless steel, length 1,5 m
Suitable for pressure up to 10 bar
Integrated flow regulator: 8 L/min per shower head
Certificazioni: EN 15154 Part 2, EN 1717