Italy
Order Entry
VWR®, Immersion Oil
VWR®, Immersion Oil
  630-1808
 :  
undefined
VWR®, Immersion Oil
  630-1808
 :  
28,60 €
  1 * 1 pezzi
  • Pk:
    1
  • Description:
    Immersion oil (8 ml)
Immersion oil is used to increase microscope resolution.
  • For immersing of the lens and specimen
  • Transparent oil with high refractive index
  • Increases the numerical aperture of an objective