Italy
Inserisci ordine
Lipophilic carbocyanine dyes and CellBrite cytoplasmic membrane labelling kits
Lipophilic carbocyanine dyes and CellBrite cytoplasmic membrane labelling kits
Supplier:  Biotium
Certificates
Lipophilic carbocyanine dyes and CellBrite cytoplasmic membrane labelling kits
Supplier:  Biotium
Product Information
  • Catalog #
    Supplier #
    Descrizione
    Conf
    Availability
    Price
  • 60010.
    60010
    DiI (DIIC18(3)), orange/red carbocyanine dye, 549/565 nm
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 202,00 €
    202,00 €
    1 * 50 mg
       202,00 €
    Supplier Article Number: 60010
    Local Article Number: BTIU60010
  • 60011.
    60011
    DiO (DIOC18(3)), green fluorescent carbocyanine dye (Ex/Em: 484/501 nm)
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 202,00 €
    202,00 €
    1 * 50 mg
       202,00 €
    Supplier Article Number: 60011
    Local Article Number: BTIU60011
  • 60012.
    60012
    DiOC14(3), derivative that is more soluble in aqueous solutions
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 202,00 €
    202,00 €
    1 * 50 mg
       202,00 €
    Supplier Article Number: 60012
    Local Article Number: BTIU60012
  • 60013.
    60013
    DIA, green fluorescent carbocyanine dye with faster diffusion in membranes than DiO
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 313,00 €
    313,00 €
    1 * 50 mg
       313,00 €
    Supplier Article Number: 60013
    Local Article Number: BTIU60013
  • 60015.
    60015
    Neuro-DIO, similar to DiO, with better solubility and faster diffusion in membranes
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 378,00 €
    378,00 €
    1 * 25 mg
       378,00 €
    Supplier Article Number: 60015
    Local Article Number: BTIU60015
  • 60016.
    60016
    Neuro-DII, similar to DiI, with faster diffusion in membranes
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 278,00 €
    278,00 €
    1 * 25 mg
       278,00 €
    Supplier Article Number: 60016
    Local Article Number: BTIU60016
  • 60018.
    60018
    DII in vegetable oil, uniformly dissolved in vegetable oil for microinjection
    0,5 mL
    Log in for availability
    Log in for availability
    1 * 0,5 mL
    1 * 0,5 mL - 301,00 €
    301,00 €
    1 * 0,5 mL
       301,00 €
    Supplier Article Number: 60018
    Local Article Number: BTIU60018
  • 60019.
    60019
    Neuro-DIO in vegetable oil, uniformly dissolved in vegetable oil for microinjection
    0,2 mL
    Log in for availability
    Log in for availability
    1 * 0,2 mL
    1 * 0,2 mL - 412,00 €
    412,00 €
    1 * 0,2 mL
       412,00 €
    Supplier Article Number: 60019
    Local Article Number: BTIU60019
  • 60020.
    60020
    Neuro-DII in vegetable oil, uniformly dissolved in vegetable oil for microinjection
    0,2 mL
    Log in for availability
    Log in for availability
    1 * 0,2 mL
    1 * 0,2 mL - 412,00 €
    412,00 €
    1 * 0,2 mL
       412,00 €
    Supplier Article Number: 60020
    Local Article Number: BTIU60020
  • 60034.
    60034
    Dilinoleyl DII (FAST DII), similar to DII, with faster diffusion in membranes
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 454,00 €
    454,00 €
    1 * 5 mg
       454,00 €
    Supplier Article Number: 60034
    Local Article Number: BTIU60034
  • 60035.
    60035
    Dilinoleyl DIO (FAST DIO), similar to DIO, with faster diffusion in membranes
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 454,00 €
    454,00 €
    1 * 5 mg
       454,00 €
    Supplier Article Number: 60035
    Local Article Number: BTIU60035
  • 60036.
    60036
    DIB, blue fluorescent carbocyanine dye (Ex/Em: 353/442 nm)
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 454,00 €
    454,00 €
    1 * 10 mg
       454,00 €
    Supplier Article Number: 60036
    Local Article Number: BTIU60036
  • 60038.
    60038
    DIOC16(3), DIO derivative used in combination with DPA for membrane potential measurements
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 545,00 €
    545,00 €
    1 * 25 mg
       545,00 €
    Supplier Article Number: 60038
    Local Article Number: BTIU60038