Stampa…
Gas purifiers, GateKeeper® Hydride
Fornitore: Entegris
ENTECE2500KFSK4REA 0 EUR
ENTECE2500KFSK4R
Gas purifiers, GateKeeper® Hydride
  • Guards against bulk gas purifier breakthrough, impurity spikes from liquid storage tanks, cylinder changeouts and unexpected contaminants in the process
  • Does not release hydrocarbons that may contaminate the process
  • Uses ambient temperature purification technology – no power or heat required
  • No moving parts