Italy
Order Entry
Flask clamps and plate holder
Flask clamps and plate holder
Supplier:  GRANT INSTRUMENTS
Certificates
undefined
Flask clamps and plate holder
Supplier:  GRANT INSTRUMENTS
Product Information
  • Catalog #
    Supplier #
    For
    Pk
    Availability
    Price
  • Flask clamp
    Flask clamp
    462-0524
    SC-25
    25 ml flasks ( TF12 holds 18, TF18 holds 33 and TF26 holds 28)
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 70,90 €
    70,90 €
    1 * 1 pezzi
       70,90 €
    Supplier Article Number: SC-25
    Local Article Number: GRANSC-25
    Specifications:
    Specifications
    Environmentally Preferable
  • Flask clamp
    Flask clamp
    462-0526
    SC-50
    50 ml flasks ( TF12 holds 14, TF18 holds 26 and TF26 holds 24)
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 61,10 €
    61,10 €
    1 * 1 pezzi
       61,10 €
    Supplier Article Number: SC-50
    Local Article Number: GRANSC-50
    Specifications:
    Specifications
    Environmentally Preferable
  • Flask clamp
    Flask clamp
    462-0522
    SC-100
    100 ml flasks ( TF12 holds 9, TF18 holds 17 and TF26 holds 15)
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 82,90 €
    82,90 €
    1 * 1 pezzi
       82,90 €
    Supplier Article Number: SC-100
    Local Article Number: GRANSC-100
    Specifications:
    Specifications
    Environmentally Preferable
  • Flask clamp
    Flask clamp
    462-0525
    SC-250
    250 ml flasks ( TF12 holds 5, TF18 holds 14 and TF26 holds 8)
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 85,10 €
    85,10 €
    1 * 1 pezzi
       85,10 €
    Supplier Article Number: SC-250
    Local Article Number: GRANSC-250
    Specifications:
    Specifications
    Environmentally Preferable
  • Flask clamp
    Flask clamp
    462-0527
    SC-500
    500 ml flasks ( TF12 holds 4, TF18 holds 6 and TF26 holds 6)
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 85,10 €
    85,10 €
    1 * 1 pezzi
       85,10 €
    Supplier Article Number: SC-500
    Local Article Number: GRANSC-500
    Specifications:
    Specifications
    Environmentally Preferable
  • Flask clamp
    Flask clamp
    462-0523
    SC-1000
    1000 ml flasks ( TF12 holds 2, TF18 holds 4 and TF26 holds 3)
    1
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 87,20 €
    87,20 €
    1 * 1 pezzi
       87,20 €
    Supplier Article Number: SC-1000
    Local Article Number: GRANSC-1000
    Specifications:
    Specifications
    Environmentally Preferable