Glass tubing, capillary
Supplier: SMITH SCIENTIFIC
Certificates
About this item
Made from Simax glass.
- Smooth, non-porous, transparent
- Catalytically indifferent
- Corrosion resistant (even in exacting operations)
- Sufficiently homogenous and without presence of heterogenous particles
Capillary tubings are used in such applications as the production of laboratory glassware, industrial apparatus, pilot plant, pipe lines, drainage, decorative glass, solar collectors and light fittings.
Product Information
- Catalog #Supplier #Tubing int.ØWall thicknessTubing ext.ØLengthPkAvailabilityPrice
- Supplier Article Number: TTC/10/2.0Local Article Number: SMITTTC/10/2.0Specifications:
Specifications
Environmentally Preferable - Supplier Article Number: TC/10/3.0Local Article Number: SMITTC/10/3.0Specifications: