Quick connect couplings, single hand, low pressure series
Supplier: NEOLAB
Certificates
About this item
One-way cordoning high-grade steel quick connect couplings.
- Temperature range: −40 to 200 °C
- Operating pressure: up to 70 bar and low vacuum
- Sealings: Viton
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- Female thread, connection G 1/8", NW 2,7 mmSupplier Article Number: 2-6305Local Article Number: NEOL2-6305Specifications:
Specifications
Environmentally Preferable - Female thread, connection M 5, NW 2,7 mmSupplier Article Number: 2-6306Local Article Number: NEOL2-6306Specifications:
Specifications
Environmentally Preferable - Female thread, connection M 5, NW 7,5 mmSupplier Article Number: 2-6308Local Article Number: NEOL2-6308Specifications:
Specifications
Environmentally Preferable - Female thread, connection G 1/8", NW 5 mmSupplier Article Number: 2-6309Local Article Number: NEOL2-6309Specifications:
Specifications
Environmentally Preferable - Plug nozzle with barbed hose connection, NW 5 mm, tube I.D. 6 mmPlug nozzle with barbed hose connection, NW 5 mm, tube I.D. 6 mm229-51252-63111Log in for availability1 * 1 Cad.Supplier Article Number: 2-6311Local Article Number: NEOL2-6311Specifications:
Specifications
Environmentally Preferable - Plug nozzle with barbed hose connection, NW 5 mm, tube I.D. 8 mmPlug nozzle with barbed hose connection, NW 5 mm, tube I.D. 8 mm229-51262-63121Log in for availability1 * 1 Cad.Supplier Article Number: 2-6312Local Article Number: NEOL2-6312Specifications:
Specifications
Environmentally Preferable - Plug nozzle with barbed hose connection, tube I.D. 8 mmPlug nozzle with barbed hose connection, tube I.D. 8 mm229-51352-63151Log in for availability1 * 1 Cad.Supplier Article Number: 2-6315Local Article Number: NEOL2-6315Specifications:
Specifications
Environmentally Preferable - Plug nozzle with barbed hose connection, tube I.D. 10 mmPlug nozzle with barbed hose connection, tube I.D. 10 mm229-51362-63161Log in for availability1 * 1 Cad.Supplier Article Number: 2-6316Local Article Number: NEOL2-6316Specifications:
Specifications
Environmentally Preferable - Plug nozzle with barbed hose connection, NW 2,7 mm, tube I.D. 4 mmPlug nozzle with barbed hose connection, NW 2,7 mm, tube I.D. 4 mm229-51152-63171Log in for availability1 * 1 Cad.Supplier Article Number: 2-6317Local Article Number: NEOL2-6317Specifications:
Specifications
Environmentally Preferable - Nipple male thread G1/8", NW 5 mm, for 2-6309Supplier Article Number: 2-6319Local Article Number: NEOL2-6319Specifications:
Specifications
Environmentally Preferable - Nipple with thread, G3/8", NW 7,8 mm, for 2-6314Supplier Article Number: 2-6322Local Article Number: NEOL2-6322Specifications: