UPLC columns, ACQUITY UPLC™ BEH C18
WATE186002352
: WATERS
- Ø int.:2,10 mm
- Particle Size:1,7 µm
- Pore size:130 Å
- Length:100 mm
- Seq:05
- Phase:C18
- Pk:1
These columns are suitable for a diverse range of analytes. With mobile phase (pH 1 to 12) and temperature (80 °C) compatibility, this trifunctionally-bonded alkyl column is a universal method development tool that can impact the retention, selectivity and sensitivity of ionisable compounds (with mobile phase pH) while delivering low- and high-pH stability for all analyte types.