Stampa…
Wipes and bottle dispenser
Fornitore: Palbam
113-8176EA 619 EUR
113-8176
Wipes and bottle dispenser
Dispenser per panni/carta
This wall-mounted dispenser is designed for wipes and IPA bottles.
  • No lid, allows one handed use
  • Two holes for stable mounting to wall
  • Stainless steel 304, electropolished finish
Used in industries such as pharmaceutical, biotechnology, medical device, microelectronics and nanotechnology - Essential to cleanroom wipe-down procedures.
Certificazioni: ISO certified
Informazioni per l'ordine: Typically available from stock for direct shipment from the supplier.