Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Italy
Products
Services
Digital Solutions
Applications
Support
About
0,00 €
Order Entry
Home
Timers
Digital Timers
Desk timer/stopwatch
Desk timer/stopwatch
Desk timer/stopwatch
609-0002
:
EA COMBS
Desk timer/stopwatch
609-0002
:
EA COMBS
1 * 1 pezzi
-
+
Colour:
Silver
Description:
Digital timer/stopwatch with jumbo digits
W×D×H:
160×70×90 mm
Pk:
1